Product Details
- SNP ID
-
rs61743570
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:109450809 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGACGGGGCTGGAGTTGGGGAAAGA[A/C]CCAGAAACAGAGCTGGACACTTACT
- Phenotype
-
MIM: 613421
MIM: 614636
- Polymorphism
- A/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
KCTD10
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs139133763] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- KCTD10
- Gene Name
- potassium channel tetramerization domain containing 10
- Gene
- MYO1H
- Gene Name
- myosin IH
There are no transcripts associated with this gene.
View Full Product Details