Product Details

SNP ID
rs190179603
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:44301354 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGACTTTGCCCTCCCCAGACCCTG[A/G]GGCAGCCCGGCTTGGCTAGCACTCA
Phenotype
MIM: 612035 MIM: 186975
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
AARS2 PubMed Links

Gene Details

Gene
AARS2
Gene Name
alanyl-tRNA synthetase 2, mitochondrial
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020745.3 Intron NP_065796.1
XM_005249245.3 Intron XP_005249302.1
XM_011514764.2 Intron XP_011513066.1
XM_017011112.1 Intron XP_016866601.1
Gene
TCTE1
Gene Name
t-complex-associated-testis-expressed 1
There are no transcripts associated with this gene.

View Full Product Details