Product Details

SNP ID
hCV25998983
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:109450724 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGGGAGGGGTAGTTTATGAGCTAT[C/G]CCTGTCACAGGGTCAGATGGCCTTC
Phenotype
MIM: 613421 MIM: 614636
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
KCTD10 PubMed Links

Gene Details

Gene
KCTD10
Gene Name
potassium channel tetramerization domain containing 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001317395.1 2098 UTR 3 NP_001304324.1
NM_001317399.1 2098 UTR 3 NP_001304328.1
NM_031954.4 2098 UTR 3 NP_114160.1
Gene
MYO1H
Gene Name
myosin IH
There are no transcripts associated with this gene.

View Full Product Details