Product Details

SNP ID
hCV25999101
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:32029498 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAAACTTTAACATCCATAAATCAC[A/C]CAGTAAAAAGTCGCTATACATAACT
Phenotype
MIM: 612032
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
DPY30 PubMed Links

Gene Details

Gene
DPY30
Gene Name
dpy-30, histone methyltransferase complex regulatory subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001321209.1 Intron NP_001308138.1
NM_001321210.1 Intron NP_001308139.1
NM_032574.3 Intron NP_115963.1

View Full Product Details