Product Details

SNP ID
rs77387357
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:19897724 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTATCAAAAGCCTAAAATAAACTCA[A/G]GGACACTTCCTTTGTTCCAAGCTCT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TTC32 PubMed Links

Gene Details

Gene
TTC32
Gene Name
tetratricopeptide repeat domain 32
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001008237.2 Intron NP_001008238.1

View Full Product Details