Product Details
- SNP ID
-
rs3124643
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
37
- Location
-
Chr.1:59133313 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GCTGGGCTGTTGCAAATGCTGTCCA[C/G]TTGCCTGGATGTTTTTCCTGGAAGG
- Phenotype
-
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
HSD52
PubMed Links
Gene Details
- Gene
- HSD52
- Gene Name
- uncharacterized LOC729467
There are no transcripts associated with this gene.
View Full Product Details