Product Details

SNP ID
rs7588069
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:236166184 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTGGAACTCCTTCTCTAGCTCCAG[C/T]AGCTGCTCGCTGGTGAAGGCAGTCC
Phenotype
MIM: 601135
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GBX2 PubMed Links

Gene Details

Gene
GBX2
Gene Name
gastrulation brain homeobox 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001301687.1 815 UTR 3 NP_001288616.1
NM_001485.3 815 Silent Mutation CTA,CTG L259L NP_001476.2
Gene
LOC105373944
Gene Name
translation initiation factor IF-2-like
There are no transcripts associated with this gene.

View Full Product Details