Product Details
- SNP ID
-
hCV25999706
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:44965107 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCCGCCACCTGGGAGAGGGCATGTA[C/G]TCAGACAGATGCTGGCCATGTGTTC
- Phenotype
-
MIM: 600642
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
MYO1G
PubMed Links
Gene Details
- Gene
- MYO1G
- Gene Name
- myosin IG
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_033054.2 |
|
Intron |
|
|
NP_149043.2 |
View Full Product Details