Product Details

SNP ID
rs12125143
Assay Type
Functionally tested
NCBI dbSNP Submissions
46
Location
Chr.1:24419715 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTACACAGTGCTCTGCATTGGCTAA[C/T]AGATATGTATGGGGTCTGCATGCCT
Phenotype
MIM: 615826
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NIPAL3 PubMed Links

Gene Details

Gene
NIPAL3
Gene Name
NIPA like domain containing 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001322854.1 Intron NP_001309783.1
NM_001322855.1 Intron NP_001309784.1
NM_001322856.1 Intron NP_001309785.1
NM_001322857.1 Intron NP_001309786.1
NM_001322858.1 Intron NP_001309787.1
NM_001322859.1 Intron NP_001309788.1
NM_001322860.1 Intron NP_001309789.1
NM_001322861.1 Intron NP_001309790.1
NM_001322862.1 Intron NP_001309791.1
NM_001322863.1 Intron NP_001309792.1
NM_001322864.1 Intron NP_001309793.1
NM_001322865.1 Intron NP_001309794.1
NM_001322866.1 Intron NP_001309795.1
NM_001322868.1 Intron NP_001309797.1
NM_020448.4 Intron NP_065181.1
XM_011541808.2 Intron XP_011540110.1
XM_017001868.1 Intron XP_016857357.1
XM_017001869.1 Intron XP_016857358.1
XM_017001870.1 Intron XP_016857359.1
Gene
STPG1
Gene Name
sperm tail PG-rich repeat containing 1
There are no transcripts associated with this gene.

View Full Product Details