Product Details

SNP ID
rs73614694
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:54662691 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTTTTCACTTGTTCAATGATGTAC[C/G]CCCAGTGTCAGGCGCTTTGCAAACA
Phenotype
MIM: 604821
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
LILRB4 PubMed Links

Gene Details

Gene
LILRB4
Gene Name
leukocyte immunoglobulin like receptor B4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278426.3 Intron NP_001265355.2
NM_001278427.3 Intron NP_001265356.2
NM_001278428.3 Intron NP_001265357.2
NM_001278429.3 Intron NP_001265358.2
NM_001278430.3 Intron NP_001265359.2
XM_017026215.1 Intron XP_016881704.1
XM_017026216.1 Intron XP_016881705.1
XM_017026217.1 Intron XP_016881706.1

View Full Product Details