Product Details

SNP ID
rs6697444
Assay Type
Functionally tested
NCBI dbSNP Submissions
27
Location
Chr.1:103533840 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCACTCCCCATGTCCCACTTCAGG[C/T]TCTGAAAAAAAAAAAAGGTATGTAG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC101928436 PubMed Links

Gene Details

Gene
LOC101928436
Gene Name
uncharacterized LOC101928436
There are no transcripts associated with this gene.

Gene
RNPC3
Gene Name
RNA binding region (RNP1, RRM) containing 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017619.3 Intron NP_060089.1

View Full Product Details