Product Details

SNP ID
rs1135321
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:177992774 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGGGAGGGGGTGGGGGGCAGGGCA[A/G]ATGGCCGGCAGGGGCTGGGTGCAAG
Phenotype
MIM: 601538
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PROP1 PubMed Links

Gene Details

Gene
PROP1
Gene Name
PROP paired-like homeobox 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006261.4 925 Silent Mutation NP_006252.3

View Full Product Details