Product Details

SNP ID
rs1140329
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:105480101 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCACACTGCCTCGCAAGCGCTCGCC[A/C]CCCTCACGTGGCTCACCTGCTGTTG
Phenotype
MIM: 123875 MIM: 601183 MIM: 603526
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
C14orf80 PubMed Links

Gene Details

Gene
C14orf80
Gene Name
chromosome 14 open reading frame 80
There are no transcripts associated with this gene.

Gene
CRIP1
Gene Name
cysteine rich protein 1
There are no transcripts associated with this gene.

Gene
CRIP2
Gene Name
cysteine rich protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001270837.1 1607 UTR 3 NP_001257766.1
NM_001270841.1 1607 UTR 3 NP_001257770.1
NM_001312.3 1607 UTR 3 NP_001303.1
Gene
MTA1
Gene Name
metastasis associated 1
There are no transcripts associated with this gene.

View Full Product Details