Product Details

SNP ID
rs12677629
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:21695198 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCTGCCTTGGTAATGATTCCAAGAT[A/T]GTCTACCCTGGTGTGGCCACCTGGA
Phenotype
MIM: 601956
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
GFRA2 PubMed Links

Gene Details

Gene
GFRA2
Gene Name
GDNF family receptor alpha 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001165038.1 Intron NP_001158510.1
NM_001165039.1 Intron NP_001158511.1
NM_001495.4 Intron NP_001486.4
XM_006716327.3 Intron XP_006716390.1
XM_011544484.2 Intron XP_011542786.1

View Full Product Details