Product Details

SNP ID
rs2670827
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:74466882 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGAATGCGGTGCGCTCTGTCTACCA[C/T]ATAAGATGGACAGATGAAGGTCCAC
Phenotype
MIM: 606790
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CD300A PubMed Links

Gene Details

Gene
CD300A
Gene Name
CD300a molecule
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256841.1 Intron NP_001243770.1
NM_007261.3 Intron NP_009192.2
XM_005256990.2 Intron XP_005257047.1
XM_005256991.4 Intron XP_005257048.1
XM_005256992.4 Intron XP_005257049.1
XM_006721656.3 Intron XP_006721719.1
XM_011524254.2 Intron XP_011522556.1

View Full Product Details