Product Details

SNP ID
rs1064527
Assay Type
Functionally tested
NCBI dbSNP Submissions
2
Location
Chr.1:109917338 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAAGAAGGCATTTCTCCTGGTACAA[G/T]ACATAATGGAGGACACCATGCGCTT
Phenotype
MIM: 120420
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CSF1 PubMed Links

Gene Details

Gene
CSF1
Gene Name
colony stimulating factor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000757.5 684 Missense Mutation GAC,TAC D91Y NP_000748.3
NM_172210.2 684 Missense Mutation GAC,TAC D91Y NP_757349.1
NM_172211.3 684 Missense Mutation GAC,TAC D91Y NP_757350.1
NM_172212.2 684 Missense Mutation GAC,TAC D91Y NP_757351.1
XM_017000369.1 684 Missense Mutation GAC,TAC D50Y XP_016855858.1

View Full Product Details