Product Details

SNP ID
rs6793239
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:48310128 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTATTTGTCAAAGACATTGAGTATA[G/T]AAAACATACTCAGTCTACAATAAAA
Phenotype
MIM: 608294
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
MIR2115 PubMed Links

Gene Details

Gene
MIR2115
Gene Name
microRNA 2115
There are no transcripts associated with this gene.

Gene
NME6
Gene Name
NME/NM23 nucleoside diphosphate kinase 6
There are no transcripts associated with this gene.

Gene
SPINK8
Gene Name
serine peptidase inhibitor, Kazal type 8 (putative)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080525.1 Intron NP_001073994.1
XM_017007046.1 Intron XP_016862535.1
XM_017007047.1 Intron XP_016862536.1

View Full Product Details