Product Details
- SNP ID
-
rs13018824
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:203333297 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ACTCATACTTTTAAATATTTTTTGT[A/G]TGCACAACTATATATCAATATTTTT
- Phenotype
-
MIM: 606442
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ABI2
PubMed Links
Gene Details
- Gene
- ABI2
- Gene Name
- abl-interactor 2
View Full Product Details