Product Details

SNP ID
rs4591150
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.16:27268260 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTGTACAAAGTAAGAAACTCCGTA[C/T]ACAAAACAGAGGGGAGCACGGAGGG
Phenotype
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
FLJ21408 PubMed Links

Gene Details

Gene
FLJ21408
Gene Name
uncharacterized LOC400512
There are no transcripts associated with this gene.

Gene
NSMCE1
Gene Name
NSE1 homolog, SMC5-SMC6 complex component
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_145080.3 144 Intron NP_659547.2
XM_006721023.3 144 UTR 5 XP_006721086.1
XM_011545761.2 144 Intron XP_011544063.1
XM_011545762.1 144 UTR 5 XP_011544064.1
XM_017023040.1 144 UTR 5 XP_016878529.1
XM_017023041.1 144 UTR 5 XP_016878530.1
XM_017023042.1 144 Intron XP_016878531.1

View Full Product Details