Product Details

SNP ID
rs4858985
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.3:88068308 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATAAATCAGTTAATTAACTTTGAGA[A/C]AGAAAGGGACAGGTTCATTTTCGGG
Phenotype
MIM: 603363
Polymorphism
A/C, Transversion Substitution
Allele Nomenclature
Literature Links
CGGBP1 PubMed Links
Additional Information
For this assay, SNP(s) [rs76951737] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
CGGBP1
Gene Name
CGG triplet repeat binding protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001008390.1 Intron NP_001008391.1
NM_001195308.1 Intron NP_001182237.1
NM_003663.3 Intron NP_003654.3
XM_017007355.1 Intron XP_016862844.1
Gene
ZNF654
Gene Name
zinc finger protein 654
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018293.2 Intron NP_060763.2
XM_011534340.2 Intron XP_011532642.2
XM_017006789.1 Intron XP_016862278.1
XM_017006790.1 Intron XP_016862279.1
XM_017006791.1 Intron XP_016862280.1
XM_017006792.1 Intron XP_016862281.1
XM_017006793.1 Intron XP_016862282.1
XM_017006794.1 Intron XP_016862283.1
XM_017006795.1 Intron XP_016862284.1

View Full Product Details