Product Details

SNP ID
rs2513279
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:68686698 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCACCACAGAGCCTGCTGGTCTCTG[C/T]GCGGCCCTCTCATCCTCTCCCTGCC
Phenotype
MIM: 137035
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GAL PubMed Links

Gene Details

Gene
GAL
Gene Name
galanin and GMAP prepropeptide
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015973.4 Intron NP_057057.2

View Full Product Details