Product Details
- SNP ID
-
rs9428576
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
71
- Location
-
Chr.1:243498725 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CATTGCCCTGTCTTGGAAATCCCCA[C/T]GTTAATGATGGCTTAAATTAGGTTG
- Phenotype
-
MIM: 611223
MIM: 613524
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
AKT3
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs9428577] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- AKT3
- Gene Name
- AKT serine/threonine kinase 3
- Gene
- SDCCAG8
- Gene Name
- serologically defined colon cancer antigen 8
View Full Product Details