Product Details
- SNP ID
-
rs4285855
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:12689541 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CAGTGCTGACTCGCGGAGAAGTGTC[C/T]CATAAATGTTAGCTGCTGCTGCTAA
- Phenotype
-
MIM: 189967
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
TEAD1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs116419326] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- TEAD1
- Gene Name
- TEA domain transcription factor 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_021961.5 |
|
Intron |
|
|
NP_068780.2 |
View Full Product Details