Product Details

SNP ID
rs216119
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:69837096 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAGCCTCATCTGACAGACAGAGAGA[C/T]GGAGACTGAGGCTCAGAAAGGCCAC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LINC00593 PubMed Links

Gene Details

Gene
LINC00593
Gene Name
long intergenic non-protein coding RNA 593
There are no transcripts associated with this gene.

View Full Product Details