Product Details

SNP ID
rs2715704
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:108823656 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAAAAACACAAAATATGTTTTATTT[C/T]CTTTCAAATTTTATTTATATACATT
Phenotype
MIM: 604962
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TRAT1 PubMed Links

Gene Details

Gene
TRAT1
Gene Name
T cell receptor associated transmembrane adaptor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001317747.1 Intron NP_001304676.1
NM_016388.3 Intron NP_057472.2

View Full Product Details