Product Details

SNP ID
rs11695176
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:101007205 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTCTACAGCTTAAGACACTTAACTA[C/T]AAGGTAAAAGAAAAGGACCAAGTAA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RPL31 PubMed Links

Gene Details

Gene
RPL31
Gene Name
ribosomal protein L31
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000993.4 4459 Intron NP_000984.1
NM_001098577.2 4459 Intron NP_001092047.1
NM_001099693.1 4459 Intron NP_001093163.1
Gene
TBC1D8
Gene Name
TBC1 domain family member 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_005263862.3 4459 UTR 3 XP_005263919.1
XM_005263863.4 4459 Intron XP_005263920.1
XM_005263864.3 4459 Intron XP_005263921.1
XM_011510507.1 4459 Intron XP_011508809.1
XM_011510508.2 4459 Intron XP_011508810.1
XM_017003198.1 4459 Intron XP_016858687.1
XM_017003199.1 4459 Intron XP_016858688.1
XM_017003200.1 4459 Intron XP_016858689.1
XM_017003201.1 4459 Intron XP_016858690.1
XM_017003202.1 4459 Intron XP_016858691.1

View Full Product Details