Product Details

SNP ID
rs4779499
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:29121475 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCCCAAGCAGGTGAAACCCCTGCGG[C/T]GTGGCCCCAGCGCACCTCAGAGACA
Phenotype
MIM: 602712
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
APBA2 PubMed Links

Gene Details

Gene
APBA2
Gene Name
amyloid beta precursor protein binding family A member 2
There are no transcripts associated with this gene.

Gene
FAM189A1
Gene Name
family with sequence similarity 189 member A1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015307.1 3641 UTR 3 NP_056122.1
XM_011521407.2 3641 UTR 3 XP_011519709.1

View Full Product Details