Product Details

SNP ID
rs11807400
Assay Type
Functionally tested
NCBI dbSNP Submissions
25
Location
Chr.1:52638413 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGTAAGCATATCTCAAATGAGAAAA[C/T]AATGTTTAATTAAAGTAACTAAAGA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FAM159A PubMed Links

Gene Details

Gene
FAM159A
Gene Name
family with sequence similarity 159 member A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001042693.2 Intron NP_001036158.1
XM_011541377.2 Intron XP_011539679.1
XM_011541378.2 Intron XP_011539680.1

View Full Product Details