Product Details

SNP ID
rs6871636
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.5:43199765 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAAGGAGAAGAGAGTTAGTAGTGAC[A/G]TTGAGTGAGCCAGGCTGCAGTGATG
Phenotype
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
NIM1K PubMed Links

Gene Details

Gene
NIM1K
Gene Name
NIM1 serine/threonine protein kinase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_153361.3 Intron NP_699192.1
XM_006714450.1 Intron XP_006714513.1
XM_006714451.1 Intron XP_006714514.1
XM_017009164.1 Intron XP_016864653.1
XM_017009165.1 Intron XP_016864654.1

View Full Product Details