Product Details

SNP ID
rs34414061
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:50669698 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGGAAAAGGGAGCAAGGCCAGGTG[C/T]TGGGATGAATAATTAAGCGAGATAA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PARP8 PubMed Links

Gene Details

Gene
PARP8
Gene Name
poly(ADP-ribose) polymerase family member 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001178055.1 Intron NP_001171526.1
NM_001178056.1 Intron NP_001171527.1
NM_024615.3 Intron NP_078891.2
XM_005248596.4 Intron XP_005248653.1
XM_011543631.2 Intron XP_011541933.1
XM_011543632.2 Intron XP_011541934.2
XM_011543633.2 Intron XP_011541935.2
XM_011543634.2 Intron XP_011541936.1
XM_011543635.2 Intron XP_011541937.1
XM_011543636.2 Intron XP_011541938.1
XM_011543640.2 Intron XP_011541942.1
XM_011543641.2 Intron XP_011541943.1
XM_011543642.2 Intron XP_011541944.1
XM_011543643.2 Intron XP_011541945.1
XM_017009850.1 Intron XP_016865339.1
XM_017009851.1 Intron XP_016865340.1

View Full Product Details