Product Details

SNP ID
rs7735614
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:73502228 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGTGGAATTCTTTCAAATGCTTAAT[G/T]TAGATTGACACATGCTACCTCTTTG
Phenotype
MIM: 602542
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
BTF3 PubMed Links

Gene Details

Gene
BTF3
Gene Name
basic transcription factor 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001037637.1 Intron NP_001032726.1
NM_001207.4 Intron NP_001198.2

View Full Product Details