Product Details

SNP ID
rs13059340
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:161344843 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATTTTGCAAAACTCAACAAATACA[C/T]GTTCAGATCTGGTTTCTCTTCAAAA
Phenotype
MIM: 610412
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SPTSSB PubMed Links

Gene Details

Gene
SPTSSB
Gene Name
serine palmitoyltransferase small subunit B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001040100.1 2255 UTR 3 NP_001035189.1
NM_001320679.1 2255 UTR 3 NP_001307608.1
XM_017005793.1 2255 UTR 3 XP_016861282.1
XM_017005794.1 2255 UTR 3 XP_016861283.1

View Full Product Details