Product Details

SNP ID
rs4381763
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:200912265 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATCATAATCTTCTCTCTCCAACCCA[G/T]ATCTTTCCTCTGGTTTCTTGTCCTA
Phenotype
MIM: 605778 MIM: 601182
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
NIF3L1 PubMed Links

Gene Details

Gene
NIF3L1
Gene Name
NGG1 interacting factor 3 like 1
There are no transcripts associated with this gene.

Gene
ORC2
Gene Name
origin recognition complex subunit 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006190.4 Intron NP_006181.1
XM_006712555.3 Intron XP_006712618.1
XM_011511252.2 Intron XP_011509554.1
XM_011511253.2 Intron XP_011509555.1

View Full Product Details