Product Details
- SNP ID
-
rs3810786
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:42326432 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- AAGTATTGAGTCTAAGATAGGGGTT[A/C]GCCTCTCTGTAGCGCCTTAATATGC
- Phenotype
-
MIM: 608150
MIM: 610750
- Polymorphism
- A/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
MIR7851
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs117901367] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- MIR7851
- Gene Name
- microRNA 7851
There are no transcripts associated with this gene.
- Gene
- PPHLN1
- Gene Name
- periphilin 1
- Gene
- ZCRB1
- Gene Name
- zinc finger CCHC-type and RNA binding motif containing 1
View Full Product Details