Product Details
- SNP ID
-
hCV26114580
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:77159749 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ACTTTCTCTTGAATATGTGACAAAA[G/T]AGAGACTACTATTTTTTAGGTATTT
- Phenotype
-
MIM: 603203
- Polymorphism
- G/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
CCNG2
PubMed Links
Gene Details
- Gene
- CCNG2
- Gene Name
- cyclin G2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_004354.2 |
|
Intron |
|
|
NP_004345.1 |
View Full Product Details