Product Details
- SNP ID
-
rs7131920
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:66305380 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTCCAGTTCCTTTCCACCTCTAAAA[C/T]TGTGATTTTAATCTCTCTTTCTAAC
- Phenotype
-
MIM: 614539
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
HELB
PubMed Links
Gene Details
- Gene
- HELB
- Gene Name
- DNA helicase B
View Full Product Details