Product Details
- SNP ID
-
rs12568730
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
24
- Location
-
Chr.1:182379152 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCTATAAAATGGCAGGGTTCTAATC[A/T]GCTCAGATTGTTTTCAGTAGAAATG
- Phenotype
-
MIM: 138290
- Polymorphism
- A/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
GLUL
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs114724499] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- GLUL
- Gene Name
- glutamate-ammonia ligase
View Full Product Details