Product Details
- SNP ID
-
rs34458044
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
36
- Location
-
Chr.1:245753113 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGATGCTGAATGCATCACCAGTAGC[A/C]AAGGGGAACAGGGGCAACCCTCAGG
- Phenotype
-
MIM: 608783
- Polymorphism
- A/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
SMYD3
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs112693109] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- SMYD3
- Gene Name
- SET and MYND domain containing 3
View Full Product Details