Product Details

SNP ID
rs34210073
Assay Type
Functionally tested
NCBI dbSNP Submissions
27
Location
Chr.1:245751488 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTTCCCTTTAGGCTGGAAGGAAAA[C/G]AGGCAGACAGAGGGGGTCTAGATTT
Phenotype
MIM: 608783
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
SMYD3 PubMed Links

Gene Details

Gene
SMYD3
Gene Name
SET and MYND domain containing 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001167740.1 Intron NP_001161212.1
NM_022743.2 Intron NP_073580.1
XM_011544253.1 Intron XP_011542555.1
XM_011544254.2 Intron XP_011542556.1
XM_011544257.1 Intron XP_011542559.1
XM_011544258.2 Intron XP_011542560.1
XM_011544259.2 Intron XP_011542561.1
XM_011544260.1 Intron XP_011542562.1
XM_011544261.2 Intron XP_011542563.1
XM_017002094.1 Intron XP_016857583.1
XM_017002095.1 Intron XP_016857584.1

View Full Product Details