Product Details

SNP ID
rs398046
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:93585725 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCCCCGGCTCCCCCACCCCGCCCG[C/G]TGCCTGCTCAGGATTGTGTCCCTGA
Phenotype
MIM: 132890
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
NR2F1 PubMed Links

Gene Details

Gene
NR2F1
Gene Name
nuclear receptor subfamily 2 group F member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005654.5 Intron NP_005645.1
XM_017009797.1 Intron XP_016865286.1
Gene
NR2F1-AS1
Gene Name
NR2F1 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details