Product Details

SNP ID
rs1080194
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:150268610 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GATGACTCACCATGCAAAAGTCAAC[C/T]AACATATAACATACTACATTAACAG
Phenotype
MIM: 606453
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LRBA PubMed Links

Gene Details

Gene
LRBA
Gene Name
LPS responsive beige-like anchor protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199282.2 Intron NP_001186211.2
NM_006726.4 Intron NP_006717.2
XM_005263372.3 Intron XP_005263429.1
XM_005263373.2 Intron XP_005263430.1
XM_005263374.3 Intron XP_005263431.1
XM_005263375.3 Intron XP_005263432.1
XM_011532434.2 Intron XP_011530736.1
XM_017008872.1 Intron XP_016864361.1
XM_017008873.1 Intron XP_016864362.1
XM_017008874.1 Intron XP_016864363.1

View Full Product Details