Product Details

SNP ID
rs4449788
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:35236783 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCAGAGGCAGTTTCAGACACAGCT[C/T]AAGTACCGGAGCTTTACCTTTGAAA
Phenotype
MIM: 616466
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
UNC5D PubMed Links

Gene Details

Gene
UNC5D
Gene Name
unc-5 netrin receptor D
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001322818.1 Intron NP_001309747.1
NM_080872.3 Intron NP_543148.2
XM_011544400.2 Intron XP_011542702.1
XM_011544401.2 Intron XP_011542703.1
XM_011544402.2 Intron XP_011542704.1
XM_011544403.2 Intron XP_011542705.1
XM_011544404.2 Intron XP_011542706.1
XM_017013044.1 Intron XP_016868533.1
XM_017013045.1 Intron XP_016868534.1
XM_017013046.1 Intron XP_016868535.1
XM_017013047.1 Intron XP_016868536.1
XM_017013048.1 Intron XP_016868537.1
XM_017013049.1 Intron XP_016868538.1
XM_017013050.1 Intron XP_016868539.1

View Full Product Details