Product Details

SNP ID
rs2332715
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:123972164 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCATGGCAACAAACTAACATAGCGG[C/G]TTTCAACCCTAAACATCATCTTTTA
Phenotype
MIM: 611757
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ROPN1 PubMed Links

Gene Details

Gene
ROPN1
Gene Name
rhophilin associated tail protein 1
There are no transcripts associated with this gene.

View Full Product Details