Product Details

SNP ID
rs4533167
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:51592960 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGATAGCAAGATCCTGTCTCTACA[A/G]CGAATAAATAAATAAAAGATAATAA
Phenotype
MIM: 610418
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
WDFY2 PubMed Links

Gene Details

Gene
WDFY2
Gene Name
WD repeat and FYVE domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_052950.3 Intron NP_443182.1
XM_011534914.1 Intron XP_011533216.1
XM_011534915.2 Intron XP_011533217.1
XM_017020386.1 Intron XP_016875875.1

View Full Product Details