Product Details

SNP ID
rs2360765
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:57471483 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTAATTCATATTAAACAACTAAAA[C/T]AGAAAAAGAAAAAAACCATATAGGA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF772 PubMed Links

Gene Details

Gene
ZNF772
Gene Name
zinc finger protein 772
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001024596.2 7156 UTR 3 NP_001019767.1
NM_001144068.1 7156 UTR 3 NP_001137540.1
XM_005258943.4 7156 UTR 3 XP_005259000.1
XM_005258944.3 7156 UTR 3 XP_005259001.1
XM_011526980.2 7156 UTR 3 XP_011525282.1
XM_017026813.1 7156 UTR 3 XP_016882302.1

View Full Product Details