Product Details

SNP ID
rs2011144
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:57659691 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGTTACCTTTGTTCTATGGGATGA[C/T]TAAAAAACTCACACACAGCTCTATA
Phenotype
MIM: 613419
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZSCAN4 PubMed Links

Gene Details

Gene
ZSCAN4
Gene Name
zinc finger and SCAN domain containing 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152677.2 230 Intron NP_689890.1
XM_011526607.2 230 Intron XP_011524909.1
XM_017026458.1 230 UTR 5 XP_016881947.1

View Full Product Details