Product Details

SNP ID
rs3862218
Assay Type
Functionally tested
NCBI dbSNP Submissions
54
Location
Chr.1:42686939 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCTGTCTCTGAAATAGACCCTAGCC[A/G]TTTTCAATTTGGTTGTTGTCCACAT
Phenotype
MIM: 606095 MIM: 154030
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PPIH PubMed Links

Gene Details

Gene
PPIH
Gene Name
peptidylprolyl isomerase H
There are no transcripts associated with this gene.

Gene
YBX1
Gene Name
Y-box binding protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004559.4 Intron NP_004550.2

View Full Product Details