Product Details

SNP ID
hCV26173191
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:81992701 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACATTGCGAATTACTGCTGATAAAT[A/C]CCCTGTTGGTTGAAAGACACTTGCC
Phenotype
MIM: 616164
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
SDR42E1 PubMed Links

Gene Details

Gene
SDR42E1
Gene Name
short chain dehydrogenase/reductase family 42E, member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_145168.2 Intron NP_660151.2

View Full Product Details