Product Details
- SNP ID
-
hCV26190831
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:27643837 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCTTCTTTACGTCTCTCCAAGGCAC[C/G]AACGTGTCTCCCTCCTGAAGCCACC
- Phenotype
-
MIM: 601879
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
LGALS9
PubMed Links
Gene Details
- Gene
- LGALS9
- Gene Name
- galectin 9
View Full Product Details