Product Details

SNP ID
rs3782681
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:8037755 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGTCATTGGAAACCTGCTCATTGGC[A/C]TGCCACTCTTCAGCTCAGATGACCC
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
FOXJ2 PubMed Links

Gene Details

Gene
FOXJ2
Gene Name
forkhead box J2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018416.2 Intron NP_060886.1
XM_011520760.2 Intron XP_011519062.1
XM_011520761.2 Intron XP_011519063.1
XM_017019640.1 Intron XP_016875129.1

View Full Product Details